Learn More
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.
£628.00 - £9175.00
Specifications
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
---|---|
Material | Polycarbonate |
Sterility | Sterile |
Temperature Range (Metric) | -100°C to 100°C |
Closure Color | White |
Product Code | Brand | Capacity (Metric) | Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Shape | Price | Quantity & Availability | ||||
16470631
|
Thermo Scientific™
CE-N3500-05 |
5 mL | Round |
£9175.00
Pack of 416 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||
16480631
|
Thermo Scientific™
CE-N3500-20 |
20 mL | Round |
£4750.00
Pack of 504 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
16420631
|
Thermo Scientific™
CE-N3120-42 |
1 L | Square |
£2290.00
Pack of 35 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
16440631
|
Thermo Scientific™
CE-N3233-42 |
2 L | Square |
£2090.00
Pack of 20 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
16450631
|
Thermo Scientific™
CE-N3405-42 |
5 L | Square |
£1050.00
Pack of 6 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||
16460631
|
Thermo Scientific™
CE-N3410-42 |
10 L | Square |
£628.00
Pack of 2 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
Description
Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
- Durable, translucent PC construction ensures the integrity of your sample
- Designed specifically for pharmaceutical and biological manufacturers
- Closures feature a silicone liner for leakproof assurance
- Extensive validation documentation support
- Excellent for freezing applications
Specifications
Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product | |
Sterile | |
White | |
Yes | |
Narrow | |
Silicone |
Polycarbonate | |
-100°C to 100°C | |
<1/3 USP <788> (Ultralow particulate) | |
Not Autoclavable | |
Plastic Media Bottle | |
Polypropylene |