Learn More
Thermo Scientific™ Nalgene™ BioTransport Carrier Handles
GREENER_CHOICE
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage.
Brand:  Thermo Scientific™ 7136-0001
92.29 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 10166252
  • £112.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage. These handles fit the BioTransport carrier for easy one-handed carrying.
  • Simple to put on and take off
  • Autoclavable stainless steel
  • Includes: Handle only. Biotransport Carrier sold separately, Cat. No. 7135-0001
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BioTransport Carrier Handle
Bio Transport Carrier Handle
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ BioTransport Carrier Handles > Handle Bio-safe Carrier