missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ BioTransport Carrier
GREENER_CHOICE

Easily view contents with the Thermo Scientific™ Nalgene™ BioTransport Carrier.

Brand:  Thermo Scientific™ 7135-0001

62.99 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.


Product Code. 10340301

  • £185.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.


Description

Description

Transport racks containing tubes or sample holders filled with potentially infectious materials using the Thermo Scientific™ Nalgene™ BioTransport Carrier, which offers excellent visibility of contents.

  • Polycarbonate carrier is tough and break resistant
  • Easy to grasp, molded-in handles
  • Silicone gasket seals tightly
  • Clamps securely hold carrier closed and assure leak-resistant seal
  • Stainless-steel handle available separately (7136-0001)
  • For accidental spills inside of carrier, autoclave with clamps closed

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Meets OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
BioTransport Carrier
Polycarbonate
1
36.8 cm
18.4 cm
17.0 cm
BioTransport Carrier
Autoclavable
4
14.5 x 7.3 x 6.687 in. (36.8 x 18.4 x 17.0cm)
14.5 in.
7.3 in.
6.687 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1284210 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number7135-0001
Lot Number1305340 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number7135-0001
Lot Number1412926 Certificate TypeCertificate of Compliance Date31/08/2024 Catalog Number7135-0001
Lot Number1398736 Certificate TypeCertificate of Compliance Date29/06/2024 Catalog Number7135-0001
Lot Number1401442 Certificate TypeCertificate of Compliance Date28/02/2024 Catalog Number7135-0001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ BioTransport Carrier > 14.5 x 7.3 x 6.75 in. (36.8 x 18.4 x 17.0cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.