Learn More
Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
Use promo code "23291" to get your promotional price.
Description
Thermo Scientific™ Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles are extremely low in trace metal content, making them an excellent choice for storing high-purity chemicals. Controlled environment manufacturing minimizes particulate contamination. Narrow-mouth bottles with certified low-particulate levels are used by shippers who design and assemble combination packaging for high-purity chemicals.
- Leakproof†
Includes:
Polypropylene closures
Recommended for:
Storage and shipping of microelectronic or biological reagents
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 8 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | Natural |
Description | Low-Particulate Narrow-Mouth Bottles |
Diameter (Metric) Inner Neck | 18 mm |
Material | HDPE |
No. per Case | 72 |
Sterility | Non-sterile |
Closure Color | Natural |
Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.