missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack Product Code.: 11379674
Add to basket

Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack
GREENER_CHOICE

Product Code. 11379674
500 mL, 28 mm
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
Closure Size:
24 mm
28 mm
38-430 mm
This item is not returnable. View return policy

Product Code. 11379674

Brand: Thermo Scientific™ 3820990500

Add to basket
Add to basket
This item is not returnable. View return policy

Effectively store high-purity chemicals with these HDPE bottles, which are extremely low in trace metal content.

Thermo Scientific™ Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles are extremely low in trace metal content, making them an excellent choice for storing high-purity chemicals. Controlled environment manufacturing minimizes particulate contamination. Narrow-mouth bottles with certified low-particulate levels are used by shippers who design and assemble combination packaging for high-purity chemicals.

Features:

  • Leakproof†

Includes:

Polypropylene closures

Recommended for:

Storage and shipping of microelectronic or biological reagents

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our 

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Natural
Description Low-Particulate Narrow-Mouth Bottles
Diameter (Metric) Inner Neck 21 mm
Material HDPE
No. per Case 48
Sterility Non-sterile
Closure Color Natural
Low Particulate Yes
Neck Finish 28 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27(c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 171 mm
Product Type Narrow Mouth Bottle
Width (English) 2.86 in.
Height (English) 6.71 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 73 mm
Closure Type Screw Closure (Assembled)
Through the Closure Dispensing No
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424580 Certificate TypeCertificate of Compliance Date14/03/2025 Catalog Number382099-0500
Lot Number1415925 Certificate TypeCertificate of Compliance Date02/11/2024 Catalog Number382099-0500
Lot Number1406832 Certificate TypeCertificate of Compliance Date06/06/2024 Catalog Number382099-0500
Lot Number1404866 Certificate TypeCertificate of Compliance Date11/04/2024 Catalog Number382099-0500
Lot Number1397502 Certificate TypeCertificate of Compliance Date08/12/2023 Catalog Number382099-0500

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.