Learn More
Thermo Scientific™ Nalgene™ HDPE Platinum Certified Clean Bottles and Carboys
Thermo Scientific™ Nalgene™ HDPE Platinum Certified Clean Bottles and Carboys are certified platinum clean and include particulate certifications to under 1/3 the allowable limits according to USP<788> and lower endotoxin levels of 0.25 EU/mL, to help mitigate potential sources of unwanted impurities.
£777.50 - £7885.00
Specifications
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
---|---|
Material | HDPE |
Sterility | Sterile |
Temperature Range (Metric) | -100°C to 120°C |
Low Particulate | <1/3 USP <788> (Ultralow particulate) |
Product Code | Brand | Capacity (Metric) | Product Type | Mouth | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Product Type | Mouth | Price | Quantity & Availability | |||
16470601
|
Thermo Scientific™
CE-E2099-0001 |
30 mL | Media Storage Bottle | Narrow |
£7885.00
Pack of 72 |
In Stock
Log in to see stock. |
|||
16470611
|
Thermo Scientific™
CE-N2099-0008 |
250 mL | Media Storage Bottle | Narrow |
£3015.00
Pack of 72 |
In Stock
Log in to see stock. |
|||
16400621
|
Thermo Scientific™
CE-N2199-0008 |
250 mL | Media Storage Bottle | Wide |
£4495.00
Pack of 72 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||
15625935
|
Thermo Scientific™
CE-N2199-0016x |
500 mL | Media Storage Bottle | Wide | N/A | ||||
16490611
|
Thermo Scientific™
CE-N2099-0032 |
1 L | Media Storage Bottle | Narrow |
£1515.00
Pack of 24 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
16420621
|
Thermo Scientific™
CE-N2199-0032 |
1 L | Media Storage Bottle | Wide |
£2870.00
Pack of 24 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||
15645935
|
Thermo Scientific™
CE-N2289-0020x |
10 L | Carboy | Narrow |
£3645.00
Pack of 6 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||
16450621
|
Thermo Scientific™
CE-N2289-0020 |
10 L | Carboy | Narrow |
£2480.00
Pack of 6 |
In Stock
Log in to see stock. |
|||
16460621
|
Thermo Scientific™
CE-N2289-0050 |
20 L | Carboy | Narrow |
£2320.00
Pack of 4 |
In Stock
Log in to see stock. |
|||
16470621
|
Thermo Scientific™
CE-N2289-0075 |
33 L | Carboy | Narrow |
£1055.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
Description
Our single-use products, including Thermo Scientific™ Nalgene™ HDPE Platinum Certified Clean Bottles and Carboys, help reduce the potential for carry-over contamination. Our new certified platinum clean products include particulate certifications to under 1/3 the allowable limits according to USP<788> and lower endotoxin levels of 0.25 EU/mL, to help mitigate potential sources of unwanted impurities.
- Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products
- Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination
- Certified low endotoxin (0.25 EU/mL) according to USP <85> to reduce pyrogens
- Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use
- Heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging
- All container/closure systems are certified leakproof* to protect valuable contents
- Robust, HDPE can be used from -100°C to 120°C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
- Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications
- Products can be supplied with forced extraction studies**, validation binders** and certificates
- Customer support includes our highly experienced commercial specialist teams, regulatory team, quality team and regional technical support
Specifications
Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product | |
Sterile | |
<1/3 USP <788> (Ultralow particulate) | |
Yes | |
Round |
HDPE | |
-100°C to 120°C | |
< 1/3 particulates allowed according to USP <778>, USP Class VI, non-cytotoxic, USP 30 <661>-compliant, non-pyrogenic, FDA 21 CFR 177.1315 | |
Not Autoclavable | |
Polypropylene |
*Our guarantee for a leakproof seal is subject to our standard product warranty, as set forth in the Thermo Fisher Scientific Terms and Conditions of Sale. Our products are leakproof at ambient temperature and pressure when used with their corresponding closures. However, to ensure safe usage, customers are advised to test our containers and closures under conditions of their planned applications. Please contact technicalsupport@thermofisher.com if you need additional information about our products.
**Forced extraction studies and validation binders from Thermo Fisher Scientific, where available, are provided under a confidentiality agreement to assist customers in product selection. Customers are responsible for determining what studies are recommended for its specific applications. Forced extraction studies and validation binders may be requested by contacting us at ROCregSupport@thermofisher.com.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.