Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
£731.00 - £2150.00
Specifications
Certifications/Compliance Comply with FDA Reg. 177.1520 and USP Class VI
Type Closed-dome Tank
Material Polypropylene
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10050461
View accessory products View Documents Promotion Details
Thermo Scientific™
2650-0020
75 L
£731.00
Each
In Stock
to see stock.
Add to basket
10338990
View accessory products View Documents
Thermo Scientific™
2650-0030
115 L
£1030.00
Each
In Stock
to see stock.
Add to basket
10469970
View accessory products View Documents
Thermo Scientific™
2650-0055
210 L
£1720.00
Each
In Stock
to see stock.
Add to basket
10595971
View Documents
Thermo Scientific™
2650-0100
378.5 L
£2150.00
Each
Estimated Shipment: 24-12-2025
to see stock.
Add to basket
Description
Description
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Warranty:
90 days
Specifications
Specifications
Comply with FDA Reg. 177.1520 and USP Class VI
Closed-dome Tank
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks