Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners
Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.
- Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
- Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Specifications
Specifications
| Type | Tank Liner |
| Material | Polyethylene |
| Capacity (English) | 100 gal. |
| Capacity (Metric) | 378 L |
| For Use With (Equipment) | Nalgene cylindrical tanks from 5 gal (19L) to 200 |
| No. per Case | 10 |
| No. per Pack | 10 |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.