missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.

Brand:  Thermo Scientific™ 343050-0055

 View more versions of this product

Product Code. 10767104

  • £752.00 / Pack of 10
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tank Liner
55 gal.
Nalgene cylindrical tanks from 5 gal (19L) to 200
10
Polyethylene
208 L
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7447900010 Certificate TypeCertificate of Processing Date13/03/2025 Catalog Number343050-0055
Lot Number7446262010 Certificate TypeCertificate of Processing Date22/01/2025 Catalog Number343050-0055
Lot Number7440310010 Certificate TypeCertificate of Processing Date22/01/2025 Catalog Number343050-0055
Lot Number7443322010 Certificate TypeCertificate of Processing Date20/01/2025 Catalog Number343050-0055
Lot Number7441284010 Certificate TypeCertificate of Processing Date20/01/2025 Catalog Number343050-0055
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners > Gamma Irradiated: 55 gal. (208L)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.