missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
48.88 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Closure Color:
Blue
Green
Red
Yellow
Description
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Color-coded top allows you to coordinate reagents with industry storage codes.
- Tip can be cut back to provide increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Yellow |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'