missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
GREENER_CHOICE

Eliminate the need for a rubber stopper with the safe-sealing solution of these continuous thread lined closures for serum vials.

Brand:  Thermo Scientific™ 342158-0023

1101.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10262523

  • £1471.92 / Pack of 2000
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Eliminate the need for a rubber stopper with the safe-sealing solution of high density polyetylene (HDPE) Thermo Scientific™ Nalgene™ Continuous Thread Lined Closures for Serum Vials.

Features:

  • Sterile to 10-6 SAL
  • Liner made of a low density polyethylene (LDPE) core between two facings of solid HDPE
  • Multiple closure colors available

Ordering Information:

Serum vial closures are bulk packed at 2,000 per case.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yellow
Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
22.1 mm
Sterile
2000
Rubber
14.2 mm
ISO 13485:2016
20-415
Serum vial closures
Yellow
HDPE
2000
Nalgene
Lined Continuous-thread Closure
2000 Cs.
Serum Vial Closure
Sterile PETG Serum Vials (Sold Separately)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7268492010 Certificate TypeCertificate of Compliance Date19/07/2017 Catalog Number342158-0023
Lot Number7261211010 Certificate TypeCertificate of Compliance Date18/07/2017 Catalog Number342158-0023
Lot Number725342310-COC Certificate TypeCertificate of Compliance Date13/04/2017 Catalog Number342158-0023
Lot Number7213517010-COC Certificate TypeCertificate of Compliance Date13/04/2017 Catalog Number342158-0023
Lot Number7257773010-COC Certificate TypeCertificate of Analysis Date13/04/2017 Catalog Number342158-0023
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack > Yellow, 20-415 closure, bulk pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.