Learn More
Thermo Scientific™ Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Eliminate the need for a rubber stopper with the safe-sealing solution of these continuous thread lined closures for serum vials.
Brand: Thermo Scientific™ 342158-0026
1101.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Eliminate the need for a rubber stopper with the safe-sealing solution of high density polyetylene (HDPE) Thermo Scientific™ Nalgene™ Continuous Thread Lined Closures for Serum Vials.
Features:
- Sterile to 10-6 SAL
- Liner made of a low density polyethylene (LDPE) core between two facings of solid HDPE
- Multiple closure colors available
Ordering Information:
Serum vial closures are bulk packed at 2,000 per case.
Specifications
Blue | |
Three-ply co-extruded liner (LDPE core between two facing of solid HDPE) | |
22.1 mm | |
Sterile | |
2000 | |
Rubber | |
14.2 mm | |
ISO 13485:2016 | |
20-415 |
Serum vial closures | |
Blue | |
HDPE | |
2000 | |
Nalgene | |
Lined Continuous-thread Closure | |
2000 Cs. | |
Serum Vial Closure | |
Sterile PETG Serum Vials (Sold Separately) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.