Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
£50.85 - £828.65
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13477649
|
Thermo Scientific™
CS509X10 |
4 in. height: 5.0mL, 81 vials per box, Case of 1 |
£50.85
Pack of 81 |
In Stock
Log in to see stock. |
|||||
12874372
|
Thermo Scientific™
CS509X24 |
2 in. height: 1.0 and 1.5mL, 100 vials per box, case of 10 |
£183.28
Pack of 10 |
In Stock
Log in to see stock. |
|||||
12745918
|
Thermo Scientific™
CS509X3 |
2 in. height: 2.0mL, 25 vials per box, case of 80 |
£828.65
Pack of 25 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
13497649
|
Thermo Scientific™
CS509X4 |
2 in. height: 2.0mL, 81 vials per box, Case of 40 |
£639.34
Pack of 40 |
In Stock
Log in to see stock. |
|||||
13487649
|
Thermo Scientific™
CS509X5 |
2 in. height: 2.0mL, 81 vials per box, Case of 20 |
£319.24
Pack of 20 |
In Stock
Log in to see stock. |
|||||
Description
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
- Warranty varies by country, please contact us for details