Learn More
Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs
Increase walk-away time and assay efficiency with these disposable polypropylene robotic reservoirs, available as a standard microplate format with 300mL volume.
£439.00 - £491.23
Specifications
Color | Natural |
---|---|
Capacity (Metric) | 300 mL |
Dimensions (L x W x H) | 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) |
Material | Polypropylene |
Type | Reservoir |
Product Code | Brand | Bottom Shape | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bottom Shape | Sterility | Price | Quantity & Availability | ||||
10466392
|
Thermo Scientific™
1200-1300 |
Flat | Non-sterile |
£439.00
Pack of 40 |
In Stock
Log in to see stock. |
||||
10723363
|
Thermo Scientific™
1200-1301 |
Flat | Sterile |
£491.23
Pack of 40 |
In Stock
Log in to see stock. |
||||
10039511
|
Thermo Scientific™
1200-2300 |
Convoluted | Non-sterile |
£450.00
Pack of 40 |
In Stock
Log in to see stock. |
||||
10373582
|
Thermo Scientific™
1200-2301 |
Convoluted | Sterile |
£491.23
Pack of 40 |
In Stock
Log in to see stock. |
||||
Description
Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.
- Convoluted bottom design is most effective with a 96-well format.
- Flat Bottom design can be used with single, six-, 12-, 96-, 384- and 1536-well formats.
- Polypropylene construction withstands robotic liquid handling, making it easier to run automated assays and increasing walk-away time.
- Reservoir is compatible with most robotic arms, plate stackers (hotels), plate cranes, and platform configurations.
- Reservoirs are RNase- & DNase-free.
- Reservoir volume is 300mL (345mL brim capacity), with molded-in graduations every 100mL.
- Reservoir is nestable for easy automated stacking and handling.
Specifications
Natural | |
3.4 x 5 x 1.75 in. (86 x 128 x 43 mm) | |
Reservoir | |
43 mm | |
86 mm | |
4 | |
128 mm |
300 mL | |
Polypropylene | |
1.75 in. | |
3.4 in. | |
40 | |
5 in. |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.