missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoTube™ Holders
GREENER_CHOICE

Product Code. p-4520556
Click to view available options
Holds:
25 Cryogenic Tubes
40 Cryogenic Tubes
50 Cryogenic Tubes
Unit Size:
Each
Pack of 15

Product Code. 10100051

Brand: Thermo Scientific™ 50300510

Only null left
Add to basket
Add to basket

Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.

Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.

  • Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
  • Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
TRUSTED_SUSTAINABILITY

Specifications

Anti-Rotation Yes
Color White
Material Polycarbonate
Type Vial Holder
For Use With (Equipment) Cryo Tube
Holds 50 Cryogenic Tubes
Product Line Nalgene
Tube Array 5 x 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.