Learn More
Thermo Scientific™ Nalgene™ Plastic Graduated Cylinder Variety Pack

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Seven sizes, from 10 to 1000mL.
Brand: Thermo Scientific™ 3662-1234
133.25 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Thermo Scientific™ Nalgene™ Plastic Graduated Cylinder Variety Packs include one each 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL cylinders. All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. No meniscus allows accurate readings every time. The large base keeps cylinders stable on the benchtop preventing tipping hazards—won't break or chip like glass labware. Suitable for food and beverage use.
- Includes seven cylinders, one each size: 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL
- Guaranteed to meet requirements of ASTM Class B, E1272 for accuracy
- No meniscus to confuse readings—eliminates guesswork
- Large blue octagonal base prevents rolling and tipping
- Generous pour spout provides ease in dispensing liquids
- Molded-in graduations are silk-screened for easy reading
- Excellent chemical resistance for a wide variety of solutions
- Impact resistant at temperatures above -40°C
- Autoclavable/Graduated
- Meets ASTM Class B, E1272 Class B and 21 CFR Part 177.1520 for food/beverage use

Specifications
10, 25, 50, 100, 250, 500, 1000 mL | |
Nalgene Grad Cylinder Variety Pack, PPCO | |
7 | |
Blue Polypropylene Base | |
121°C |
|
Round | |
Food and Beverage Use, Refreshing an Existing Lab | |
Graduated Cylinder Set |
PPCO | |
ASTM, CFR 21 Part 177.1520, E1272 | |
mL | |
Autoclavable | |
Yes | |
250°F | |
Octagonal |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1290574 | Certificate of Compliance | 10/12/2024 | 3662-1234 |
1304808 | Certificate of Compliance | 10/12/2024 | 3662-1234 |
1296932 | Certificate of Compliance | 10/12/2024 | 3662-1234 |
1414897 | Certificate of Compliance | 26/09/2024 | 3662-1234 |
1410727 | Certificate of Compliance | 27/08/2024 | 3662-1234 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Notes:
- Autoclaving will affect graduation accuracy