missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Griffin Low-Form Plastic Beaker Variety Pack
37.21 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Capacity (Metric):
50 mL, 100 mL, 150 mL, 250 mL, 400 mL, 600 mL, and 1000 mL
Description
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Excellent chemical resistance to a wide variety of lab reagents, easy to read silk screened graduations and convenient stacking capabilities make these beakers an excellent choice for daily lab activities. Thermo Scientific™ Nalgene™ Griffin Low-Form Beaker Variety Pack includes one each 50mL, 100mL, 150mL, 250mL, 400mL, 600mL and 1000mL PPCO beakers.
- Includes seven beakers, one each size: 50mL, 100mL, 150mL, 250mL, 400mL, 600mL and 1000mL
- No drip single-spout design provides precision pouring control
- With ribs for easy stacking
- Silk screened with graduations, size code, resin code, catalog number, max/ temperature and “No Flame” symbol for easy identification and safe use
- Impact resistant at temperatures above -40°C
- Autoclavable/Graduated
- High density polyethylene
Compliance:
- Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Beakers and 21 CFR Part 177.1520 for food/beverage use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Natural translucent |
Description | Griffin Low-Form Beaker Variety Pk. |
Capacity (Metric) | 50 mL, 100 mL, 150 mL, 250 mL, 400 mL, 600 mL, and 1000 mL |
Material | PPCO |
Quantity | 1/Cs. |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Certifications/Compliance | ISO/DIN 7056, 21 CFR Part 177.15 |
Scale | mL |
Show More |
Notes:
- Autoclaving will affect graduation accuracy.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'