missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Low-Profile Carboy

For Use With: Dispense most liquids, including reagents, distilled water and titrants.; Closure Material: Polypropylene; Color: White

Brand:  Thermo Scientific™ 2323-0008

431.10 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.



Additional Details : Weight : 5.48960kg

 View more versions of this product

Product Code. 15941491

  • £571.00 / Pack of 4
Estimated Shipment: 25-10-2024
to see stock.
Add to basket

Description

Description

Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with most aqueous and alcohol lab liquids, including reagents, distilled water and titrants. Convenient integral carry handle for safe, easy transport. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:
  • Stack up to three high to save space on benchtop or in refrigerator
  • Contour handle allows easy one-handed carrying—even when full
  • Excellent chemical resistance—can tolerate strong acids and bases for a wide variety of solutions
  • Leakproof† construction means more secure samples

Includes:

Spigot and spigot closure and four acid-proof, pressure-sensitive labels

Note:

These tests, using other liquids, may not yield the same results. To ensure safe usage, Nalgene advises customers to test bottles and closures under conditions of their planned applications. Nalgene bottles and carboys are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Specifications

Not Autoclavable
8 L
White
HDPE
2 gal.
Polypropylene
Dispense most liquids, including reagents, distilled water and titrants.
Low-profile Carboy
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.