missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Large HDPE Funnels
Choose a safer alternative to glass with this shatterproof funnel, featuring external and internal ribbing for air release.
Brand: Thermo Scientific™ 4262-0110
52.03 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
Description
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
- High-density polyethylene is shatterproof, making it a reliable long-term tool for the lab
Specifications
258mm | |
25 mm | |
301 mm | |
Funnel Large HDPE 279mm | |
1/Pk.; 6/Cs. |
4.06 L | |
HDPE | |
Not Autoclavable | |
90 mm |