Learn More
Thermo Scientific™ Nalgene™ Large HDPE Funnels
Choose a safer alternative to glass with this shatterproof funnel, featuring external and internal ribbing for air release.
Brand: Thermo Scientific™ 4262-0080
37.98 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 55
Additional Details : Weight : 0.32850kg
Description
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
- High-density polyethylene is shatterproof, making it a reliable long-term tool for the lab
Specifications
201mm | |
27 mm | |
246 mm | |
Funnel Large HDPE 203mm | |
1/Pk.; 6/Cs. |
1.9 L | |
HDPE | |
No | |
88 mm |