Learn More
Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure

Description
Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.
- 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
- Lighter weight than glass alternatives make for easy lifting
- Translucence allows for easy viewing of liquid levels
- Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
- Autoclavable/Leakproof†
Includes:
Polypropylene screw caps
Note:
Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 0.5 gal., 2 L |
Capacity (Metric) | 2 L |
Capacity (English) | 0.5 gal. |
Color | Translucent |
Description | Large Narrow-Mouth Bottle PP 2L |
Diameter (Metric) Inner Neck | 25 mm |
Material | PPCO |
No. per Case | 6 |
No. per Pack | 1 |
Sterility | Non-sterile |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1418784 | Certificate of Compliance | 12/12/2024 | 2203-0005 |
1357167 | Certificate of Compliance | 10/12/2024 | 2203-0005 |
1298675 | Certificate of Compliance | 10/12/2024 | 2203-0005 |
1411442 | Certificate of Compliance | 16/08/2024 | 2203-0005 |
1407088 | Certificate of Compliance | 14/06/2024 | 2203-0005 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.