Learn More
Thermo Scientific™ Nalgene™ Large Polypropylene Waste Containers with Cover
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with these large polypropylene waste containers featuring secure, friction-fit closures.
Specifications
| Certifications/Compliance | Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogensAutoclave upright with lid on. |
|---|---|
| Autoclavable | Autoclavable |
| Quantity | 4/Cs. |
Description
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with Thermo Scientific™ Nalgene™ Large Polypropylene Waste Containers with Cover. These containers feature a secure, friction-fit closure and molded-in handgrips. They meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
- Molded-in handgrips make lifting easy
- Autoclavable/Biohazard
Note:
Autoclave this container upright with the lid on.
.
Specifications
| Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogensAutoclave upright with lid on. | |
| Autoclavable | |
| 4/Cs. |