Learn More
Thermo Scientific™ Nalgene™ LDPE Centrifuge Bottle Adapter
Support 250mL bottles and ensure proper rotor fit with the Thermo ScientificTM NalgeneTM LDPE Centrifuge Bottle Adapter.
Brand: Thermo Scientific™ DS3125-0250
56.60 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Support 250mL bottles and ensure proper rotor fit with the Thermo Scientific™ Nalgene™ LDPE Centrifuge Bottle Adapter. For use with Thermo Scientific™ Sorvall™ GSA rotors..
- For use with Nalgene 250ml centrifuge bottles catalog numbers 3120-0250, 3121-0250, 3122-0250, 3127-0250, 3140-0250 and 3141-0250
- Adapts Nalgene 250ml centrifuge bottles to fit in Thermo Scientific Sorval GSA rotors
- Molded of white, low-density polyethylene
Specifications
Adapter | |
LDPE | |
Centrifuge Bottle Adapter for 250mL bottles | |
6 |
250 mL Bottle | |
250 mL | |
White |