Learn More
Thermo Scientific™ Nalgene™ LDPE, Round Carboy
Description
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.
- Molded-in graduations of 5 L and 1 gal. increments
- Leakproof†
Includes:White polypropylene screw cap and 83B closure
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (English) | 6.6 gal. |
| Capacity (Metric) | 25 L |
| Closure Material | Polypropylene |
| Closure Size | 83B |
| Color | White |
| Diameter (Metric) Inner Neck | 64 mm |
| Diameter (Metric) Outer | 287 mm |
| For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
| Height (Metric) With Closure | 594 mm |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.