Learn More
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
£239.22 - £259.87
Product Code | Brand | Holds | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | For Use With (Equipment) | Price | Quantity & Availability | ||||
10542144
|
Thermo Scientific™
HR509X19A |
Cryogenic Box, 81-Ampule | Locator Jr. |
£239.22
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
12787065
|
Thermo Scientific™
HR509X20A-70 |
Cryogenic Box, 81-Ampule | For: Locator 4 Plus |
£259.87
Each |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||||
10185433
|
Thermo Scientific™
HR509X19A-70 |
Cryogenic Box, 81-Ampule | For Locator Jr. Plus |
£243.52
Each |
In Stock
Log in to see stock. |
||||
Description
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
- Designed for long-term storage
- Features an indexed frame and box organizational system
- Unique hanging rack promotes easy sample retrieval
- Suited for both manual and computerized inventory record-keeping methods
Specifications
Cryogenic Box, 81-Ampule | |
Locator™ Plus Cryogenic Rack |