Learn More
Thermo Scientific™ Nalgene™ Magnetic PVDF Carboy Stirrer
Beschreibung
Mix high volumes of media and buffer solution at low speed using the Thermo Scientific™ Nalgene™ Magnetic PVDF Carboy Stirrer. Adjusts easily to fit either a 10 or 20L Nalgene carboy with an 83B size closure. Supplied with a closure and two impellers. Eliminates the need to retrieve magnets from your solution.
- For use with a magnetic stir plate (not included)
- Includes closure and two impellers
- Autoclavable
Spezifikation
Spezifikation
| Closure Size | 83B |
| Description | Stir bar D x L: 1/2 x 3 in. |
| For Use With (Equipment) | 10 or 20L Nalgene carboys with 83B closure |
| Length (English) | 3 in. |
| Length (Metric) | 588 mm |
| Material | PTFE, PVDF, Stainless Steel |
| No. per Case | 1 Ea. |
| No. per Pack | 1 Ea. |
| Product Type | Stirrer |
| Quantity | Each |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.