Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
GREENER_CHOICE
Store, ship and package liquids with these highly reliable, lab-quality bottles, specifically designed for long-term demanding lab use.
£22.55 - £119.00
Specifications
Color Translucent
Description Lab Quality Narrow-Mouth Bottle
Material HDPE
Sterility Non-sterile
Closure Color Natural
View More Specs
Products 11
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10718331
GREENER_CHOICE
View Documents
Thermo Scientific™
2002-9125
4 mL 13 mm
£22.55
Pack of 12
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
10550022
GREENER_CHOICE
View Documents
Thermo Scientific™
2002-9025
8 mL 20 mm
£23.05
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10758711
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-9050
15 mL 20 mm
£24.80
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10768711
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-0001
30 mL 20 mm
£28.60
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10313501
GREENER_CHOICE
View Documents
Thermo Scientific™
2002-0002
60 mL 20 mm
£34.20
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10159140
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-0004
125 mL 24 mm
£38.95
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10782711
GREENER_CHOICE
View Documents
Thermo Scientific™
2002-0006
175 mL 24 mm
£52.80
Pack of 12
In Stock
to see stock.
Add to basket
10507341
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-0008
250 mL 24 mm
£53.75
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10749671
GREENER_CHOICE
View Documents
Thermo Scientific™
2002-9016
500 mL 38-430 mm
£119.00
Pack of 12
In Stock
to see stock.
Add to basket
10517341
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-0016
500 mL 28 mm
£81.65
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10027950
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2002-0032
1 L 38-430 mm
£60.00
Pack of 6
In Stock
to see stock.
Edge
Add to basket
Description
Description
Store, ship, and package liquids using highly reliable and durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
  • Versatile and indispensable in the lab
  • Uniform walls provide durability and resistance to splitting or punctures
  • Can be used in a freezer up to -100°C (-149°F)
  • Linerless polypropylene closures ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
Specifications
Translucent
HDPE
Natural
Yes
Round
Polypropylene
No
Lab Quality Narrow-Mouth Bottle
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Narrow
Unlined
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.