Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Packaging Bottles with Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE
Minimize biological and particulate contamination with sterile packaging bottles produced in a controlled environment and featuring chemical resistance.
£172.00 - £1205.00
Specifications
Color Translucent
Description Narrow-Mouth Packaging Bottles
Material HDPE
Sterility Sterile
Closure Color White
View More Specs
Products 6
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10304103
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0001
30 mL 20-415 mm
£1205.00
Pack of 864
In Stock
to see stock.
Add to basket
10621294
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0002
60 mL 20-415 mm
£863.00
Pack of 540
In Stock
to see stock.
Edge
Add to basket
10190173
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0004
125 mL 24-415 mm
£465.00
Pack of 240
In Stock
to see stock.
Add to basket
10474743
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0008
250 mL 24-415 mm
£279.00
Pack of 180
In Stock
to see stock.
Add to basket
10262903
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0016
500 mL 28-415 mm
£509.00
Pack of 120
In Stock
to see stock.
Add to basket
10465123
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
342089-0032
1000 mL 38-430 mm
£172.00
Pack of 24
In Stock
to see stock.
Add to basket
Description
Description
Minimize biological and particulate contamination with Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Packaging Bottles with Closure. With durable construction and a narrow mouth for easy pouring, these high density polyethylene bottles are produced in a controlled environment, feature excellent chemical resistance and are sterile to 10-6 SAL.
Features:
  • Produced in a controlled environment 
  • Natural high-density polyethylene bottles with white, linerless polypropylene closures
  • Sterile to 10-6 SAL
  • RNAse, DNAse, hDNA and pyrogen free
  • Leakproof† when used with Nalgene closures
Ordering Information:
Bottles are packed in shrink-wrapped trays with closures assembled.
Specifications
Specifications
Translucent
HDPE
White
Certified: Sterile (SAL 10-6), RNAse, DNAse, hDNA and pyrogen free. Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Narrow
Sterile
Polypropylene
Narrow-Mouth Packaging Bottles
Sterile
Yes
Yes
Round
Packer Bottle
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Packaging Bottles with Closure: Sterile, Shrink-Wrapped Trays