missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure Product Code.: 11349614

Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure

Product Code. 11349614
1 L, 38-430 mm
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20 mm
24 mm
28 mm
38-430 mm
This item is not returnable. View return policy

Product Code. 11349614

Brand: Thermo Scientific™ 20870032

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth PPCO economy bottles with closure.

Perform light-duty, general-purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure. These polypropylene copolymer bottles are lightweight versions of the original Nalgene Lab Quality bottle. Made from high quality laboratory grade plastic materials for reliably low leachables and extractables.

Features:
  • Economical and recyclable; designed for single use
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof protection without the use of a liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof†
Note:
  • Completely disengage threads or remove cap before autoclaving
Includes:
  • Polypropylene closures
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1 L
Capacity (English) 32 oz.
Color Translucent
Description Narrow-Mouth Economy Bottle
Diameter (Metric) Inner Neck 27 mm
Material PPCO
No. per Case 24
No. per Pack 24
Sterility Non-sterile
Closure Color Natural
Neck Finish 38 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Autoclavable Autoclavable
Disposable Single-use
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 216 mm
Product Type Narrow Mouth Bottle
Height (English) 8.5 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 91 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1303389 Certificate TypeCertificate of Compliance Date23/11/2020 Catalog Number2087-0032
Lot Number1272153 Certificate TypeCertificate of Compliance Date02/01/2020 Catalog Number2087-0032
Lot Number1267690 Certificate TypeCertificate of Compliance Date07/11/2019 Catalog Number2087-0032
Lot Number1263272 Certificate TypeCertificate of Compliance Date11/09/2019 Catalog Number2087-0032
Lot Number1260807 Certificate TypeCertificate of Compliance Date29/07/2019 Catalog Number2087-0032

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.