missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
Chemical Label:
Ethyl Alcohol
Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, but contents stay in bottle with no risk of leakage
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Natural |
Certifications/Compliance | Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'