Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
£34.24
Specifications
Description | RTK Wash Bottle |
---|---|
Material | LDPE |
Certifications/Compliance | Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. |
Closure Included | Yes |
Graduated | No |
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
10562123
|
Thermo Scientific™
2425-0502 |
500 mL | Ethyl Alcohol | N/A | |||||
Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, so contents stay in bottle; prevents self-dispensing
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
.
Specifications
RTK Wash Bottle | |
Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. | |
No | |
Cylindrical | |
Screw Cap |
LDPE | |
Yes | |
Narrow | |
Safety Wash Bottle | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.