missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack Product Code.: 10630914
Add to basket

Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack
GREENER_CHOICE

Product Code. 10630914
15 mL, 20-415 mm
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
This item is not returnable. View return policy

Product Code. 10630914

Brand: Thermo Scientific™ 3120069050

Add to basket
Add to basket
This item is not returnable. View return policy

Sample, store and ship small-volume, valuable reagents with these autoclavable bottles that can be used in test kits and many small-volume applications.

Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable, and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Linerless closure, included, ensures performance without wrinkles that can cause leaks or contamination of your solutions.

Features:
  • Nonsterile
  • Autoclavable

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 oz., 15 mL
Capacity (Metric) 15 mL
Capacity (English) 0.5 oz.
Color Translucent
Diameter (Metric) Inner Neck 14 mm
Material PPCO
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 58 mm
Product Type Sample Container
Height (English) 2.3 in. (With Closure); 2.2 in. (Without Closure)
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 25 mm
Closure Type Screw Cap
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.