Learn More
Thermo Scientific™ Nalgene™ Non-Phthalate PVC Tubing
GREENER_CHOICE
Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65 standards
£29.15 - £280.00
Specifications
Certifications/Compliance Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Description Non-Phthalate PVC Tubing
Autoclavable Autoclavable
Material PVC
Sterility Non-sterile
View More Specs
Products 25
Product Code Brand Diameter (English) Inner Diameter (English) Outer Overall Length (English) Maximum Pressure Price Quantity & Availability  
Product Code Brand Diameter (English) Inner Diameter (English) Outer Overall Length (English) Maximum Pressure Price Quantity & Availability  
16585130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-9020
1/8 in. 1/4 in. 250 ft. 50 psig at 73°F (23°C), 25 psig at 125°F (51°C)
£122.57
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16595130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-9030
3/16 in. 5/16 in. 250 ft. 36 psig at 73°F (23°C), 18 psig at 125°F (51°C)
£156.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
16555130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-4060
1/4 in. 3/8 in. 100 ft. 30 psig at 73°F (23°C), 15 psig at 125°F (51°C)
£135.00
Each
In Stock
to see stock.
Edge
Add to basket
16505140
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-9090
5/16 in. 7/16 in. 250 ft. 28 psig at 73°F (23°C), 14 psig at 125°F (51°C)
£236.90
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16565130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-4120
3/8 in. 1/2 in. 100 ft. 22 psig at 73°F (23°C), 11 psig at 125°F (51°C)
£131.00
Each
In Stock
to see stock.
Add to basket
16575130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-4140
3/8 in. 5/8 in. 100 ft. 40 psig at 73°F (23°C), 20 psig at 125°F (51°C)
£238.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
16585120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0160
7/16 in. 5/8 in. 50 ft. 30 psig at 73°F (23°C), 15 psig at 125°F (51°C)
£97.20
Each
Estimated Shipment: 13-08-2025
to see stock.
Add to basket
16595120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0180
1/2 in. 5/8 in. 50 ft. 20 psig at 73°F (23°C), 10 psig at 125°F (51°C)
£83.30
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
16505130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0190
1/2 in. 11/16 in. 50 ft. 26 psig at 73°F (23°C), 13 psig at 125°F (51°C)
£119.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
16515130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0200
1/2 in. 3/4 in. 50 ft. 30 psig at 73°F (23°C), 15 psig at 125°F (51°C)
£164.00
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16525130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0260
5/8 in. 7/8 in. 50 ft. 26 psig at 73°F (23°C), 13 psig at 125°F (51°C)
£187.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
16535130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0310
3/4 in. 1 in. 50 ft. 22 psig at 73°F (23°C), 11 psig at 125°F (51°C)
£280.00
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16545130
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0350
1 in. 1 1/4 in. 50 ft. 18 psig at 73°F (23°C), 14 psig at 125°F (51°C)
£260.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
16565110
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0010
1/8 in. 3/16 in. 50 ft. 30 psig at 73°F (23°C), 15 psig at 125°F (51°C)
£29.15
Each
In Stock
to see stock.
Add to basket
16575110
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0020
1/8 in. 1/4 in. 50 ft. 50 psig at 73°F (23°C), 25 psig at 125°F (51°C)
£40.65
Each
In Stock
to see stock.
Add to basket
16585110
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0030
3/16 in. 5/16 in. 50 ft. 36 psig at 73°F (23°C), 18 psig at 125°F (51°C)
£40.15
Each
In Stock
to see stock.
Add to basket
16595110
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0060
1/4 in. 3/8 in. 50 ft. 30 psig at 73°F (23°C), 15 psig at 125°F (51°C)
£68.15
Each
In Stock
to see stock.
Add to basket
16505120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0070
1/4 in. 7/16 in. 50 ft. 40 psig at 73°F (23°C), 20 psig at 125°F (51°C)
£84.40
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16515120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0080
1/4 in. 1/2 in. 50 ft. 46 psig at 73°F (23°C), 23 psig at 125°F (51°C)
£100.54
Each
In Stock
to see stock.
Add to basket
16525120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0090
5/16 in. 7/16 in. 50 ft. 28 psig at 73°F (23°C), 14 psig at 125°F (51°C)
£77.65
Each
In Stock
to see stock.
Add to basket
16535120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0100
5/16 in. 1/2 in. 50 ft. 36 psig at 73°F (23°C), 18 psig at 125°F (51°C)
£85.55
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16545120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0110
5/16 in. 9/16 in. 50 ft. 40 psig at 73°F (23°C), 20 psig at 125°F (51°C)
£107.00
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16555120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0120
3/8 in. 1/2 in. 50 ft. 22 psig at 73°F (23°C), 11 psig at 125°F (51°C)
£71.24
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16565120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0130
3/8 in. 9/16 in. 50 ft. 32 psig at 73°F (23°C), 16 psig at 125°F (51°C)
£118.00
Each
In Stock
to see stock.
Add to basket
16575120
GREENER_CHOICE
View Documents
Thermo Scientific™
8701-0140
3/8 in. 5/8 in. 50 ft. 40 psig at 73°F (23°C), 20 psig at 125°F (51°C)
£133.12
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Nalgene™ Non-Phthalate Clear Plastic PVC Tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene 180 tubing but is free from orthophthalate DEHP plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and complies with California proposition 65. Resists hardening and discoloration from a wide range of laboratory chemicals and does not impart odor or taste making this tubing ideal for a wide range of non-medical, general lab, research and pharmaceutical applications.
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Free from ortho phthalate plasticizers
  • Resists hardening and discoloration from a wide range of chemicals - demonstrates dimensional stability
  • Equivalent to Tygon™ E-3603 tubing
  • Marked with I.D. and regulatory compliance as well as at 12 in. intervals for easy identification and measurement (except as noted below)
  • Durometer hardness is Shore A, 60
  • Does not impart odor or taste
  • Can write on with Nalgene Lab marker
  • Slips easily over glass and plastic fittings; grips securely
  • Autoclavable/Transparent
Please recycle the corrugate cartons in which your Nalgene  products are packaged.
Size 8701-0010 is not printed.
Compliance
FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65
Specifications
Specifications
Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Autoclavable
Non-sterile
Clear
4 x OD bend radius
160°F
Flexible Tubing
Non-Phthalate PVC Tubing
PVC
Autoclave, Gas, Chemical
60 (Shore A)
Very low
71°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Non-Phthalate PVC Tubing