Learn More
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.
£160.00 - £467.00
Specifications
Closure Material | Polypropylene, red rubber |
---|---|
Material | Polycarbonate |
No. per Case | 50 |
No. per Pack | 10 |
Sterility | Non-sterile |
Alert:
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
10402121
![]() |
Thermo Scientific™
3138-0030 |
30 mL | 102.6 mm | 25.8 mm |
£202.00
Pack of 10 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||
10088800
![]() |
Thermo Scientific™
3138-0010 |
10 mL | 83.8 mm | 16.1 mm |
£177.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
10088810
![]() |
Thermo Scientific™
3138-0050 |
50 mL | 114.8 mm | 28.8 mm |
£467.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
10179900
![]() |
Thermo Scientific™
3138-0016 |
16 mL | 107.2 mm | 18.2 mm |
£160.00
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||
Description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.
Features:- Withstand temperatures from -135° to +135°C (-211° to +275°F)
- Round bottom to fit high speed rotors
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
- Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
- Autoclavable; transparent; Leakproof when used with supplied Nalgene closure†; Repeated autoclaving will reduce the life of PC centrifuge tubes.
Ordering Information:
Replacement sealing cap assemblies available for 30ml and 50ml sizes. Use Cat. No. DS3132-0020 for 30mL tube and Cat. No. DS3132-0024 for 50mL tube.
Notes:
- Repeated autoclaving will shorten the life of polycarbonate centrifuge tubes.
- Must be filled to at least 80% total capacity for proper performance.
- Consult the Thermo Scientific Centrifugeware Chemical Resistance Table for chemical compatibility ratings.
- Cat. Nos. 3138-0050 is designed to fit universal rotors. Actual brim capacity of 3138-0050 is 42mL.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Polypropylene, red rubber | |
50 | |
Non-sterile | |
Round | |
Screw Cap | |
No | |
50,000 x g |
Polycarbonate | |
10 | |
Autoclavable | |
Round | |
No | |
Centrifuge Tube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.