Learn More
Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter

Easily remove the Thermo ScientificTM NalgeneTM Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use.
Brand: Thermo Scientific™ DS3147-0050
57.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Easily remove the Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use. This autoclavable adapter must be used with Nalgene 35L conical Oak Ridge Tubes (Cat. Nos. 3146, 3148).
Ordering Information:
To assure leakproof service, especially at speeds greater than 10,000 x g or when spinning hazardous materials, use Nalgene sealing cap assemblies DS3131-0020 or -0024.
Includes:
Supplied with linerless, polypropylene screw closure which will not leak during ordinary use.
Requires:
Both tubes require white polycarbonate tube adapter (see DS3147 0050 below) to centrifuge.

Specifications
35 mL | |
6 | |
Autoclavable | |
Conical | |
50,000 x g |
Polycarbonate | |
6 | |
Conical | |
Centrifuge Tube Adapter |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1351634 | Certificate of Compliance | 10/12/2024 | DS3147-0050 |
1121191 | Certificate of Compliance | 19/07/2017 | DS3147-0050 |
1077462-COC | Certificate of Compliance | 17/04/2017 | DS3147-0050 |
1065277-COC | Certificate of Compliance | 17/04/2017 | DS3147-0050 |
1020858-COC | Certificate of Compliance | 17/04/2017 | DS3147-0050 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.