Learn More
Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles
Description
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
- Durable, crystal clear PETG for sample integrity
- Closures with polyethylene foam core liners for leakproof† assurance
- Custom options available for fluid transfer applications
- Supported by extensive validation documentation
- Suitable for storage in conditions from -40° to +70°C
Warranty: 90 days

Specifications
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Clear |
Diameter (English) | 3 in. |
Diameter (English) Inner Neck | 1.1 in. |
Diameter (Metric) Inner Neck | 28 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Material | PETG |
No. per Case | 70/Cs. |
Packaging | Bulk |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Scientific Resources
Brochures
Product Information
Schematics & Diagrams
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1420621 | Certificate of Processing | 03/02/2025 | 3005-70,300570 |
1360828 | Certificate of Processing | 04/12/2024 | 3005-70 |
1332840 | Certificate of Processing | 04/12/2024 | 3005-70 |
1420621 | Certificate of Compliance | 24/01/2025 | 3005-70,300570 |
1387594 | Certificate of Compliance | 10/12/2024 | 3005-70 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.