missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ PETG Certified Clean Containers Product Code.: 16460581
Add to basket

Thermo Scientific™ Nalgene™ PETG Certified Clean Containers

Product Code. 16460581
500 mL
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
This item is not returnable. View return policy

Product Code. 16460581

Brand: Thermo Scientific™ 3820190500

Add to basket
Add to basket
This item is not returnable. View return policy

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers.

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.

Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.

  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.

Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Diameter (English) 2.9 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Material PETG
No. per Case 24/Cs.
No. per Pack 4/Pk.
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Low Particulate USP <788> (Low particulate)
Certifications/Compliance USP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), non-cytotoxic, non-pyrogenic, and non-hemolytic.
Closure Included Yes
Autoclavable Not Autoclavable
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 177 mm
Product Type Clean Container
Height (English) 6.97 in.
Closure Size 38-430 mm
Diameter (Metric) 74 mm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1421412 Certificate TypeCertificate of Processing Date10/02/2025 Catalog Number382019-0500
Lot Number1420398 Certificate TypeCertificate of Processing Date20/01/2025 Catalog Number382019-0500
Lot Number1421412 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number382019-0500
Lot Number1420398 Certificate TypeCertificate of Compliance Date09/01/2025 Catalog Number382019-0500
Lot Number1416002 Certificate TypeCertificate of Compliance Date27/12/2024 Catalog Number382019-0500

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ PETG Certified Clean Containers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.