missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Ultra-Low Particulate Vials

Product Code. 16450611
Click to view available options
Capacity (Metric):
5 mL
20 mL
Unit Size:
Pack of 416
Pack of 504
This item is not returnable. View return policy

Product Code. 16450611

Brand: Thermo Scientific™ CEN20350005

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for the most critical applications, Nalgene PETG Ultra-Low Particulate Vials are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • Leakproof High Density Polyethylene (HDPE) closures
  • Serum Vials are packed in shrink-wrapped trays with closures attached
  • Manufactured in compliance with ISO 13485:2003 quality requirements
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
No. per Case 416/Cs.
No. per Pack 52/Pk.
Quantity 5 mL
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance ISO 13485:2003, 21 CFR 177.1520, USP Class VI
Product Type Media Bottle
Workflow Step Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.