missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle Product Code.: 16460611
Add to basket

Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle

Product Code. 16460611
20 mL
Click to view available options
Capacity (Metric):
5 mL
20 mL
This item is not returnable. View return policy

Product Code. 16460611

Brand: Thermo Scientific™ CEN20350020

Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • Leakproof High Density Polyethylene (HDPE) closures
  • Serum Vials are packed in shrink-wrapped trays with closures attached
  • Manufactured in compliance with ISO 13485:2003 quality requirements
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 20 mL
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
No. per Case 504/Cs.
No. per Pack 84/Pk.
Quantity Case of 504
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance ISO 13485:2003, 21 CFR 177.1520, USP Class VI
Product Type Media Bottle

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7308455010 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog NumberCE-N2035-0020
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.