Learn More
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile

Description
Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
- Sterile to 10-6 SAL
- Non-pyrogenic and non-cytotoxic
- 20-415 lined white high-density polyethylene closure attached to PETG vial
- Leakproof†
Ordering Information:
Serum Vials are packed in shrink-wrapped trays with closures attached.
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 0.16 oz., 5 mL |
Capacity (Metric) | 5 mL |
Capacity (English) | 0.16 oz. |
Diameter (Metric) Inner Neck | 11 mm |
Material | PETG |
No. per Case | 100 |
No. per Pack | 20 |
Sterility | Sterile |
Neck Finish | 20-415 mm |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1308974 | Certificate of Processing | 04/12/2024 | 2035-0005 |
1321928 | Certificate of Processing | 04/12/2024 | 2035-0005 |
1352835 | Certificate of Processing | 04/12/2024 | 2035-0005 |
1328984 | Certificate of Processing | 04/12/2024 | 2035-0005 |
1299019 | Certificate of Processing | 04/12/2024 | 2035-0005 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
79Hours
0Minutes
9Seconds
52New Offer Save Up to 85%
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.