missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile Product Code.: 10704043
Add to basket

Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
GREENER_CHOICE

Product Code. 10704043
10 mL
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
This item is not returnable. View return policy

Product Code. 10704043

Brand: Thermo Scientific™ 20350010

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that are leakproof when used with provided Nalgene lined white HDPE closures.

Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.

Features:
  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • 20-415 lined white high-density polyethylene closure attached to PETG vial
  • Leakproof†

Ordering Information:

Serum Vials are packed in shrink-wrapped trays with closures attached.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.33 oz., 10 mL
Capacity (Metric) 10 mL
Capacity (English) 0.33 oz.
Color White
Diameter (Metric) Inner Neck 11 mm
Material PETG
No. per Case 100
No. per Pack 20
Sterility Sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 56 mm
Product Type Sample Container
Height (English) 2.2 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 24 mm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424997 Certificate TypeCertificate of Compliance Date21/03/2025 Catalog Number2035-0010
Lot Number1419820 Certificate TypeCertificate of Compliance Date19/02/2025 Catalog Number2035-0010
Lot Number1414632 Certificate TypeCertificate of Processing Date11/03/2025 Catalog Number2035-0010
Lot Number1412936 Certificate TypeCertificate of Processing Date11/03/2025 Catalog Number2035-0010
Lot Number1387854 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number2035-0010

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.