missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
GREENER_CHOICE

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Brand:  Thermo Scientific™ 322032-0005

558.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10282853

  • £703.60 / Pack of 1656
Estimated Shipment: 21-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Vials
Clear
PETG
5 mL
276
Nalgene
Screw Cap (Not Included)
276 Pk. , 1656 Cases
Serum Vial
Lined Continuous Thread Closure
11 mm
22 mm
Non-sterile
1656
1/6 oz.
5 mL
44 mm
ISO 13485:2003
20-415
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers