missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Brand:  Thermo Scientific™ 322032-0010

Code : NEW

Additional Details : Weight : 9.97900kg

 View more versions of this product

Product Code. 10513765

  • £570.50 / Pack of 1260
Estimated Shipment: 17-04-2025
to see stock.
Add to basket

Description

Description

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

Specifications

Specifications

Serum Vials
Clear
PETG
10 mL
252
Nalgene
Screw Cap (Not Included)
252 Pk., 1260 Cases
Serum Vial
Lined Continuous Thread Closure
11 mm
24 mm
Non-sterile
1260
1/3 oz.
10 mL
54 mm
ISO 13485:2003
20-415
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers