Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.
£355.21 - £703.60
Specifications
Description | Serum Vials |
---|---|
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Material | PETG |
Sterility | Non-sterile |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10282853
![]() |
Thermo Scientific™
322032-0005 |
5 mL |
£703.60
Pack of 1656 |
Estimated Shipment: 29-12-2025
Log in to see stock. |
||||||
10513765
![]() |
Thermo Scientific™
322032-0010 |
10 mL |
£575.82
Pack of 1260 |
Estimated Shipment: 29-12-2025
Log in to see stock. |
||||||
10406663
![]() |
Thermo Scientific™
322032-0020 |
20 mL |
£355.21
Pack of 612 |
In Stock
Log in to see stock. |
||||||
Description
Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).
- PETG
- Non-cytotoxic
- Non-pyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Nonsterile
- Leakproof† with Nalgene lined continuous thread closures (available separately)
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).
Specifications
Serum Vials | |
Clear | |
Non-sterile | |
ISO 13485:2003 | |
20-415 |
11 mm | |
PETG | |
Nalgene | |
Serum Vial | |
Lined Continuous Thread Closure |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.