Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
£371.39 - £1020.00
Specifications
Description | Serum Vials |
---|---|
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Material | PETG |
Sterility | Sterile |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10455123
![]() |
Thermo Scientific™
342032-0005 |
5 mL |
£1020.00
Pack of 1656 |
In Stock
Log in to see stock. |
||||||
10740005
![]() |
Thermo Scientific™
342032-0010 |
10 mL |
£602.23
Pack of 1260 |
In Stock
Log in to see stock. |
||||||
10600334
![]() |
Thermo Scientific™
342032-0020 |
20 mL |
£371.39
Pack of 612 |
In Stock
Log in to see stock. |
||||||
Description
Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).
- PETG
- Non-cytotoxic
- Non-pyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
- Leakproof† with Nalgene lined continuous thread closures (Cat. No. 342158)
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).
Specifications
Serum Vials | |
Clear | |
Sterile | |
Lined Continuous-thread Closure (Sold Separately) | |
Serum Vial | |
Lined Continuous-Thread Closure |
11 mm | |
PETG | |
Nalgene | |
ISO 13485:2016 | |
20-415 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.