missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.
Brand: Thermo Scientific™ 342030-0010
528.74 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.
Features:
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
Ordering Information:
Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.
Specifications
Serum Vials | |
Clear | |
PETG | |
10 mL | |
252 | |
Nalgene | |
Stoppers and Caps (Not Included) | |
1260 Cases, 252 Pk. | |
Serum Vial | |
Stopper, Cap |
13 mm | |
24 mm | |
Sterile | |
1260 | |
1/3 oz. | |
10 mL | |
50 mm | |
ISO 13485:2016 | |
20 mm |