Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.
Brand: Thermo Scientific™ 342030-0003
882.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.
Features:
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
Ordering Information:
Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.
Specifications
Serum Vials | |
Clear | |
PETG | |
3 mL | |
493 | |
Nalgene | |
Stoppers and Caps (Not Included) | |
3451 Cases, 493 Pk. | |
Serum Vial | |
Stopper, Cap |
7 mm | |
17 mm | |
Sterile | |
3451 | |
1/10 oz. | |
3 mL | |
37 mm | |
ISO 13485:2016 | |
13 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.