missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays

Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:
- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile
10-6
SAL - Molded in graduations
- Closure supplied separately
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 1 oz., 30 mL |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Clear |
Description | Media bottles without closure |
Diameter (English) | 1.5 in. |
Diameter (English) Inner Neck | 0.55 in. |
Diameter (Metric) Inner Neck | 14 mm |
For Use With (Application) | Filling Lines |
Material | PETG |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1421414 | Certificate of Processing | 03/02/2025 | 342024-0030 |
1374273 | Certificate of Processing | 04/12/2024 | 342024-0030 |
1394939 | Certificate of Processing | 04/12/2024 | 342024-0030 |
1383853 | Certificate of Processing | 04/12/2024 | 342024-0030 |
1421414 | Certificate of Compliance | 23/01/2025 | 342024-0030 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
86Hours
9Minutes
21Seconds
53New Offer Save Up to 85%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays >
Spot an opportunity for improvement?Share a Content Correction