Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Brand: Thermo Scientific™ 4109-0125
47.90 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
Specifications
4 oz. | |
125 mL | |
24 | |
Autoclavable | |
Yes | |
Non-sterile | |
Screw |
Flask Erlenmeyer Screw Closure PMP 125mL | |
PMP | |
6 | |
Reusable | |
Narrow | |
Plain | |
33 mm |