Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Marke: Thermo Scientific™ 4109-0250
66.15 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 55
Weitere Details : Weight : 0.17910kg
Beschreibung
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
Spezifikation
8 oz. | |
250 mL | |
12 | |
Autoclavable | |
Yes | |
Non-sterile | |
Screw |
Flask Erlenmeyer Screw Closure PMP 250mL | |
PMP | |
4 | |
Reusable | |
Narrow | |
Plain | |
38 mm |